Få mere at vide
Glycolic-2,2-d2 Acid, CDN
Artikelnummer. 30308748
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308748 0.25 g
30308943 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308748 Leverandør CDN Leverandørnr. CDND55340.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycolic-2,2-d2 acid
CAS 75502-10-2
Molekylær formel C2 D2 H2 O3
Mængde 0.25 g
InChI formel InChI=1 S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)/i1D2
SMIL [2 H]C([2 H])(O)C(=O)O
IUPAC navn 2,2-dideuterio-2-hydroxy-acetic acid
Molekylvægt (g/mol) 78.06
Formel vægt 78.0286 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity