Få mere at vide
Glycitein, MedChemExpress
Artikelnummer. 16461268
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16461268 5 mg
16408828 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16461268 Leverandør Medchem Express Leverandørnr. HYN0016
MedChemExpress Glycitein is a soybean (yellow cultivar) isoflavonoid; used in combination with other isoflavonoids such as genistein and daidzein to study apoptosis and anti-oxidation processes.
Glycitein is a soybean (yellow cultivar) isoflavonoid
Tekniske data
Kemisk navn eller materiale Glycitein
CAS 40957-83-3
Farve White
Renhedsgrad noter Research
Molekylær formel C16H12O5
Mængde 5 mg
Synonym Glycetein
Opløselighedsinformation DMSO : 25 mg/mL (87.95 mM; Need ultrasonic)
SMIL O=C(C(C1=CC=C(O)C=C1)=CO2)C3=C2C=C(O)C(OC)=C3
Molekylvægt (g/mol) 284.26
Formel vægt 284.26
Procent renhed 98.17%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death