Få mere at vide
Glycinexylidide (2-Amino-N-(2,6-dimethylphenyl)acetamide), Mikromol™
Artikelnummer. 17964006
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17964006 100 mg
17970596 25 mg
30459645 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17964006 Leverandør Mikromol Leverandørnr. MM0102.17
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Glycinexylidide (2-Amino-N-(2,6-dimethylphenyl)acetamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycinexylidide
CAS 18865-38-8
Analyt- eller komponentnavne Glycinexylidide
Forsendelsestilstand Room Temperature
Molekylær formel C10 H14 N2 O
Certificeringer/compliance ISO 17025
Synonym Acetamide, 2-amino-N-(2,6-dimethylphenyl)-, 2',6'-Acetoxylidide, 2-amino- (6CI,8CI), 2-Amino-N-(2,6-dimethylphenyl)acetamide, 2-Amino-2',6'-dimethylacetanilide, Astra 6204, GX, Glycine xylidide, Glycyl xylidide, N-Glycyl-2,6-xylidine, W 49167, Ω-Amino-2,6-dimethylacetanilide, omega-Amino-2,6-dimethylacetanilide
InChI formel InChI=1S/C10H14N2O/c1-7-4-3-5-8(2)10(7)12-9(13)6-11/h3-5H,6,11H2,1-2H3,(H,12,13)
SMIL Cc1cccc(C)c1NC(=O)CN
IUPAC navn 2-amino-N-(2,6-dimethylphenyl)acetamide
Molekylvægt (g/mol) 178.23
Mængde 100 mg
Formel vægt 178.1106 g/mol