Få mere at vide
Glycine-N,N,O-d3, CDN
Artikelnummer. 30309559
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30309559 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309559 Leverandør CDN Leverandørnr. CDND50861G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycine-N,N,O-D3
CAS 4896-76-8
Molekylær formel C2 D3 H2 N O2
Mængde 1 g
Synonym Glycine-N,N,1-d3 (6 CI, 7 CI, 8 CI, 9 CI), Glycine-N,N,O-d3, Glycine-N,N,O-D3
InChI formel InChI=1 S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i/hD3
SMIL [2 H]OC(=O)CN([2 H])[2 H]
IUPAC navn deuterio 2-(dideuterioamino)acetate
Molekylvægt (g/mol) 78.0851
Formel vægt 78.0509 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity