missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ACS Grade, ≥98.5%, Thermo Scientific Chemicals

Catalog Number p-8361382
Click to view available options
Quantity:
2.5 kg
25 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
This item is not returnable. View return policy
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Melting Point ∼245°C (decomposition)
Linear Formula H2NCH2COOH
Beilstein 635782
Packaging Plastic Bottle
Formula Weight 75.07
Grade ACS
Density 1.595
Quantity 2.5 kg
Specific Gravity 1.595
Physical Form Crystalline Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'