Få mere at vide
Glycine-2,2-d2, CDN
Artikelnummer. 30312549
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30312549 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312549 Leverandør CDN Leverandørnr. CDND00645G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycine-d2
CAS 4896-75-7
Molekylær formel C2 2H2 H3 N O2
Mængde 5 g
InChI formel InChI=1 S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1D2
SMIL [2 H]C([2 H])(N)C(=O)O
IUPAC navn 2-amino-2,2-dideuterioacetic acid
Molekylvægt (g/mol) 77.08
Formel vægt 77.0446 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity