Få mere at vide
Glycidyl Stearate-d35, TRC
Artikelnummer. 30240698
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240698 5 mg
30240474 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240698 Leverandør Toronto Research Chemicals Leverandørnr. TRCG6159875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246820-60-9
Molekylær formel C21H5D35O3
Molekylvægt (g/mol) 375.76
Synonym Octadecanoic Acid-d35 2-Oxiranylmethyl Ester, Stearic Acid-d35 2,3-Epoxypropyl Ester, Glycidyl Octadecanoate-d35, NSC 404228-d35, Stearic Acid-d35 Glycidyl Ester;
SMIL O=C(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])OCC1OC1
Tekniske data
Kemisk navn eller materiale Glycidyl Stearate-d35
Mængde 5 mg
InChI formel InChI=1S/C21H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h20H,2-19H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2
Formel vægt 375.5174