Få mere at vide
Glycidyl Palmitate-D5, TRC
Artikelnummer. 30403580
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30403580 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403580 Leverandør Toronto Research Chemicals Leverandørnr. TRCG61595410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycidyl Palmitate-d5
CAS 1794941-80-2
Renhedsgrad noter HPLC
Molekylær formel C19 D5 H31 O3
Mængde 10 mg
Synonym [Dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate, Glycidyl palmitate-d5, Hexadecanoic acid (2-oxiranyl-1,3,3-d3)methyl-d2 ester, Palmitic acid 2,3-epoxypropyl-1,1,2,3,3-d5 ester, (±)-Glycidyl-d5 palmitate, Glycidyl-d5 hexadecanoate, Glycidyl palmitate D5 (glycidyl-D5), Palmitic acid glycidyl-d5 ester
InChI formel InChI=1S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3/i16D2,17D2,18D
SMIL [2H]C([2H])(OC(=O)CCCCCCCCCCCCCCC)C1([2H])OC1([2H])[2H]
IUPAC navn [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate
Molekylvægt (g/mol) 317.518
Formel vægt 317.298
Procent renhed >95