missing translation for 'onlineSavingsMsg'
Få mere at vide

Glycidyl Palmitate-d5 (1.0 mg/mL in Acetonitrile), TRC

Product Code. 30484223
Change view
Click to view available options
Mængde:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30484223 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30484223 Supplier Toronto Research Chemicals Supplier No. TRCKIT97971X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Glycidyl Palmitate-d5
CAS 1794941-80-2
Molekylær formel C19 D5 H31 O3
Mængde 1 mL
Synonym [Dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate, Glycidyl palmitate-d5, Hexadecanoic acid (2-oxiranyl-1,3,3-d3)methyl-d2 ester, Palmitic acid 2,3-epoxypropyl-1,1,2,3,3-d5 ester, (±)-Glycidyl-d5 palmitate, Glycidyl-d5 hexadecanoate, Glycidyl palmitate D5 (glycidyl-D5), Palmitic acid glycidyl-d5 ester
InChI formel InChI=1S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3/i16D2,17D2,18D
SMIL [2H]C([2H])(OC(=O)CCCCCCCCCCCCCCC)C1([2H])OC1([2H])[2H]
IUPAC navn [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate
Molekylvægt (g/mol) 317.518
Formel vægt 317.298
Koncentration eller sammensætning (efter analyt eller komponenter) Glycidyl Palmitate-d5 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.