Få mere at vide
Glycidyl Palmitate-d31, TRC
Artikelnummer. 30407767
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30407767 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407767 Leverandør Toronto Research Chemicals Leverandørnr. TRCG6159522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycidyl Palmitate-d31
CAS 1246819-24-8
Molekylær formel C19H5D31O3
Mængde 2.5 mg
Synonym Hexadecanoic Acid-d31 2-Oxiranylmethyl Ester, Palmitic Acid-d31 2,3-Epoxypropyl Ester, (+/-)-Glycidyl Palmitate-d31, Glycidyl Hexadecanoate-d31, NSC 406558-d31, Palmitic Acid-d31 Glycidyl Ester
InChI formel InChI=1S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2
SMIL O=C(OCC1OC1)C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]
IUPAC navn oxiran-2-ylmethyl hexadecanoate-d31
Molekylvægt (g/mol) 343.68
Formel vægt 343.46