Få mere at vide
Glycidylmethacrylat, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2,3-Epoxypropyl methacrylate |
| CAS | 106-91-2 |
| Molekylær formel | C7 H10 O3 |
| Mængde | 250 mg |
| Synonym | 2-Propenoic acid, 2-methyl-, 2-oxiranylmethyl ester, 2-Propenoic acid, 2-methyl-, oxiranylmethyl ester (9CI), Methacrylic acid, 2,3-epoxypropyl ester (6CI,7CI,8CI), (±)-Glycidyl methacrylate, 2-Methylacrylic acid oxiranylmethyl ester, 2-[(Methacryloyloxy)methyl]oxirane, 3-Methacryloyloxy-1,2-epoxypropane, Acryester G, Acryester GMA, Acryester GU, Blemmer G, Blemmer GH, Blemmer GH-LC, Blemmer GMA, Blemmer GP, Blemmer GS, EM 54, Epoxypropyl methacrylate, Glycidol methacrylate, Glycidyl methacrylate, Glycidyl α-methylacrylate, Light Ester G, M 0590, Methacryloyloxymethyloxirane, NSC 24156, NSC 67195, P 1901, SR 379, SY-Monomer G, Sartomer 379, Sartomer SR 379 |
| InChI formel | InChI=1S/C7H10O3/c1-5(2)7(8)10-4-6-3-9-6/h6H,1,3-4H2,2H3 |
| SMIL | CC(=C)C(=O)OCC1CO1 |
| IUPAC navn | oxiran-2-ylmethyl 2-methylprop-2-enoat |
| Molekylvægt (g/mol) | 142.15 |
| Formel vægt | 142.063 |