Få mere at vide
Glycidyl Laurate-d5 (1.0 mg/mL in isopropanol), TRC
Artikelnummer. 30482780
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30482780 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482780 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT83771X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycidyl Laurate-d5
CAS 1329563-35-0
Molekylær formel C15H23D5O3
Mængde 1 mL
Synonym Dodecanoic Acid 2-Oxiranylmethyl-d5 Ester, Lauric Acid 2,3-Epoxypropyl-d5 Ester, 2,3-Epoxy-1-propanol-d5 Laurate, Glycidyl-d5 Dodecanoate, Lauric Acid Glycidyl-d5 Ester, NSC 406541-d5
InChI formel InChI=1S/C15H28O3/c1-2-3-4-5-6-7-8-9-10-11-15(16)18-13-14-12-17-14/h14H,2-13H2,1H3/i12D2,13D2,14D
SMIL [2H]C1(C(O1)([2H])C([2H])([2H])OC(=O)CCCCCCCCCCC)[2H]
IUPAC navn [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] dodecanoate
Molekylvægt (g/mol) 261.41
Formel vægt 261.2352
Koncentration eller sammensætning (efter analyt eller komponenter) Glycidyl Laurate-d5 1 mg/mL