Få mere at vide
Glyceryl Tri(octadecanoate-18,18,18-d3), CDN
Artikelnummer. 30319210
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319210 0.1 g 0.1g
30311540 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319210 Leverandør CDN Leverandørnr. CDND53610.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tristearin-D9 (tri(octadecanoyl-18,18,18-D3))
CAS 285979-76-2
Molekylær formel C57 D9 H101 O6
Mængde 0.1 g
Synonym Octadecanoic-18,18,18-d3 acid, 1,2,3-propanetriyl ester (9 CI), Octadecanoic-18,18,18-d3 acid 1,2,3-propanetriyl ester, Glyceryl Tri(octadecanoate-18,18,18-d3), Glycerol trioctadecanoate-d9, Glyceryl tristearate-d9, Trioctadecanoin-d9, Tristearin-d9, Tristearin-D9 (tri(octadecanoyl-18,18,18-D3))
InChI formel InChI=1 S/C57H110O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h54H,4-53H2,1-3H3/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC([2 H])([2 H])[2 H])OC(=O)CCCCCCCCCCCCCCCCC([2 H])([2 H])[2 H]
IUPAC navn 2,3-bis(18,18,18-trideuteriooctadecanoyloxy)propyl 18,18,18-trideuteriooctadecanoate
Molekylvægt (g/mol) 900.5352
Formel vægt 899.8867 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity