Få mere at vide
Glyceryl Tri(hexadecanoate-2,2-d2), CDN
Artikelnummer. 30320955
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320955 0.25 g -
30309493 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320955 Leverandør CDN Leverandørnr. CDND53580.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tripalmitin-D6 (tri(hexadecanoyl-2,2-D2))
CAS 241157-06-2
Molekylær formel C51 D6 H92 O6
Mængde 0.25 g
Synonym Hexadecanoic-2,2-d2 acid, 1,2,3-propanetriyl ester (9 CI), Hexadecanoic-2,2-d2 acid 1,2,3-propanetriyl ester, Glyceryl Tri(hexadecanoate-2,2-d2), Glycerol trihexadecanoate-2,2,2',2',2'',2''-d6, Glyceryl tripalmitate-d6, Trihexadecanoin-d6, Tripalmitin-d6, Tripalmitin-D6 (tri(hexadecanoyl-2,2-D2))
InChI formel InChI=1 S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i43D2,44D2,45D2
SMIL [2 H]C([2 H])(CCCCCCCCCCCCCC)C(=O)OCC(COC(=O)C([2 H])([2 H])CCCCCCCCCCCCCC)OC(=O)C([2 H])([2 H])CCCCCCCCCCCCCC
IUPAC navn 2,3-bis(2,2-dideuteriohexadecanoyloxy)propyl 2,2-dideuteriohexadecanoate
Molekylvægt (g/mol) 813.3572
Formel vægt 812.774 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity