Få mere at vide
Glyceryl 1,3-Distearate-d5, TRC
Artikelnummer. 30406055
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30406055 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406055 Leverandør Toronto Research Chemicals Leverandørnr. TRCG60149225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glyceryl 1,3-Distearate-d5
CAS 1246523-69-2
Molekylær formel C39H71D5O5
Mængde 25 mg
Synonym Octadecanoic Acid 2-Hydroxy-1,3-Propanediyl Ester-d5, 1,3-di-Stearin-d5, 1,3-Di-O-stearoylglycerol-d5, 1,3-Distearin-d5, 1,3-Distearoylglycerin-d5, 1,3-Distearoylglycerol-d5, Glycerin 1,3-Distearate-d5, Glycerol 1,3-Distearate-d5, Glyceryl 1,3-Distearate-d5, NSC 404229-d5, Stearic Acid Diglycerin Ester-d5, sn-1,3-Distearoylglycerol-d5
InChI formel InChI=1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3/i35D2,36D2,37D
SMIL [2H]C([2H])(OC(=O)CCCCCCCCCCCCCCCCC)C([2H])(O)C([2H])([2H])OC(=O)CCCCCCCCCCCCCCCCC
IUPAC navn (1,1,2,3,3-pentadeuterio-2-hydroxy-3-octadecanoyloxypropyl) octadecanoate
Molekylvægt (g/mol) 630.05
Formel vægt 629.6007