Få mere at vide
(±)-Glyceryl-1,1,2,3,3-d5 1-Monooleate, CDN
Artikelnummer. 30325723
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325723 0.1 g
30310495 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325723 Leverandør CDN Leverandørnr. CDND76100.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Oleoyl Glycerol-d5
CAS 565183-24-6
Molekylær formel C21 D5 H35 O4
Mængde 0.1 g
Synonym 9-Octadecenoic acid (9 Z)-, 2,3-dihydroxypropyl-1,1,2,3,3-d5 ester (9 CI, ACI), 2,3-Dihydroxypropyl-1,1,2,3,3-d5 (9 Z)-9-octadecenoate (ACI), (±)-Glyceryl-1,1,2,3,3-d5 1-Monooleate, (9 Z)-9-Octadecenoic acid 2,3-dihydroxypropyl-1,1,2,3,3-d5 ester, α-Glyceryl-d5 monooloeate, α-Monoolein-d5, Glyceryl 1-Monooleate-D5 (glyceryl-D5)
InChI formel InChI=1 S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,22-23 H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20 D
SMIL [2 H]C([2 H])(O)C([2 H])(O)C([2 H])([2 H])OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC navn (1,1,2,3,3-pentadeuterio-2,3-dihydroxypropyl) (Z)-octadec-9-enoate
Molekylvægt (g/mol) 361.5707
Formel vægt 361.324 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity