Få mere at vide
Glycerol-d8, TRC
Artikelnummer. 30405614
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30405614 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405614 Leverandør Toronto Research Chemicals Leverandørnr. TRCG179326250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycerol-d8
CAS 7325-17-9
Molekylær formel C3 D8 O3
Mængde 250 mg
Synonym 1,2,3-Propane-1,1,2,3,3-d5-triol-1,2,3-d3 (ACI), 1,2,3-Propane-1,1,2,3,3-d5-triol-d3 (9CI), Glycerol-d8 (7CI, 8CI), 1,1,2,3,3-Pentadeuterio-1,2,3-trideuteriooxypropane, Glycerol-D8
InChI formel InChI=1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1D2,2D2,3D,4D,5D,6D
SMIL [2H]OC([2H])([2H])C([2H])(O[2H])C([2H])([2H])O[2H]
IUPAC navn 1,1,2,3,3-pentadeuterio-1,2,3-trideuteriooxypropane
Molekylvægt (g/mol) 100.1431
Formel vægt 100.0976