missing translation for 'onlineSavingsMsg'
Få mere at vide

Glycerol-d5, TRC

Artikelnummer. 30389768
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30389768 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389768 Supplier Toronto Research Chemicals Supplier No. TRCG598402100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Glycerol-d5
CAS 62502-71-0
Molekylær formel C3 2H5 H3 O3
Mængde 100 mg
Synonym Glycerol-d5, 1,2,3-Propane-1,1,2,3,3-d5-triol
InChI formel InChI=1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1D2,2D2,3D
SMIL [2H]C([2H])(O)C([2H])(O)C([2H])([2H])O
IUPAC navn 1,1,2,3,3-pentadeuteriopropane-1,2,3-triol
Molekylvægt (g/mol) 97.12
Formel vægt 97.0787
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.