missing translation for 'onlineSavingsMsg'
Få mere at vide

Glycerol 1-Monobutyrate (Technical Grade), TRC

Product Code. 17981945
Change view
Click to view available options
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17981945 100 mg
17984625 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17981945 Supplier Toronto Research Chemicals Supplier No. TRCG598415100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-25-5
Molekylær formel C7H14O4
Molekylvægt (g/mol) 162.18
Synonym 1-Monobutyrin, 2,3-Dihydroxypropyl Butyrate, Glycerol 1-Monobutyrate, Glycerol α-Monobutyrate, Glycerol-α-mono-n-butyrate, Glyceryl 3-Monobutyrate NSC 8451, α-Monobutyrin
IUPAC navn 2,3-dihydroxypropyl butanoate
SMIL CCCC(=O)OCC(O)CO

Specifications

Kemisk navn eller materiale Glycerol 1-Monobutyrate
Mængde 100 mg
InChI formel InChI=1S/C7H14O4/c1-2-3-7(10)11-5-6(9)4-8/h6,8-9H,2-5H2,1H3
Formel vægt 162.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.