Få mere at vide
Glycerol-1,1,2,3,3-d5, CDN
Artikelnummer. 30315934
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315934 5 g
30309223 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315934 Leverandør CDN Leverandørnr. CDND13875G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycerol-d5
CAS 62502-71-0
Molekylær formel C3 2H5 H3 O3
Mængde 5 g
InChI formel InChI=1 S/C3H8O3/c4-1-3(6)2-5/h3-6 H,1-2H2/i1D2,2D2,3 D
SMIL [2 H]C([2 H])(O)C([2 H])(O)C([2 H])([2 H])O
IUPAC navn 1,1,2,3,3-pentadeuteriopropane-1,2,3-triol
Molekylvægt (g/mol) 97.12
Formel vægt 97.0787 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity