Få mere at vide
Glycerin Rosin Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Glycerin Rosin Ester |
| CAS | 8050-31-5 |
| Mængde | 1 g |
| Synonym | 138A, 138AL, 140A, 140A (Ester), 8BG, 90I, Abiester 90, Bevilite 62-88, Bevipale 85, Bremasin 1380, DM 399010G, DS 70L, Dertoline G 2L, Dertoline G 5, Dertoline RC, Dertoline RC 2, Dertopoline CG, E 445, E 445 (Rosin Ester), Eastman Ester Gum 8D, Eastman Ester Gum 8L-M, Ester Gum 105, Ester Gum 8BG, Ester Gum AAG, Ester Gum AAV, Ester Gum RE, Ester Gum S 100, Fuda 90I, Gedir 100M, Glycerides, Rosin, Glycerol Ester Of Disproportionated Rosin, Glycerol Ester Of Wood Rosin, Granolite SG, Hariester DS 70L, Hariester DS 816, Hariester DS 90, Hariester DS 90S, Hariester L, Hariester S, Hariester T, Hariester TF, Hercules Ester Gum 8D, K-Solution, K-Solution NK, MG 905, Neotall 85, Permalyn 2095, Permalyn 5095, Pinova Ester Gum 8BG, R 2100 (Resin), Resiester GUM 35V, Resinall 625, Rosin Glyceride, Rosin, Glycerol Ester, S 202, S 202 (Tackifier), S 780G, Snowtack 780G, Snowtack 790, Stepilac R 80, Super Ester A 18, Super Ester S 100, Sylvalite RE 85L, Vinsol Ester Gum, Wood Rosin, Ester With 1,2,3-Propanetriol, Zonester 55 |