Få mere at vide
Glyburide-d3 (methoxy-d3), TRC
Artikelnummer. 30406934
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30406934 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406934 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59835125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glyburide-d3 (methoxy-d3)
CAS 1219803-02-7
Renhedsgrad noter HPLC
Molekylær formel C23 D3 H25 Cl N3 O5 S
Mængde 25 mg
Synonym Benzamide, 5-chloro-N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-(methoxy-d3)- (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-(methoxy-d3)benzamide (ACI), Glibenclamide-d3, Glibenclamide-D3 (methoxy-D3), Glyburide-d3 (methoxy-d3)
InChI formel InChI=1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)/i1D3
SMIL [2H]C([2H])([2H])Oc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC navn 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-(trideuteriomethoxy)benzamide
Molekylvægt (g/mol) 497.022
Formel vægt 496.1626
Procent renhed >95