Få mere at vide
Glyburide-d3 (methoxy-d3), CDN
Artikelnummer. 30328335
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328335 0.01 g
30321735 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328335 Leverandør CDN Leverandørnr. CDND60110.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glyburide-d3 (methoxy-d3)
CAS 1219803-02-7
Molekylær formel C23 D3 H25 Cl N3 O5 S
Mængde 0.01 g
Synonym Benzamide, 5-chloro-N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-(methoxy-d3)- (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-(methoxy-d3)benzamide (ACI), Glibenclamide-d3, Glibenclamide-D3 (methoxy-D3), Glyburide-d3 (methoxy-d3)
InChI formel InChI=1 S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18 H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)/i1D3
SMIL [2 H]C([2 H])([2 H])Oc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC navn 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-(trideuteriomethoxy)benzamide
Molekylvægt (g/mol) 497.022
Formel vægt 496.1626 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity