Få mere at vide
Glyburide-d11, TRC
Artikelnummer. 30404321
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30404321 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404321 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59835210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glyburide-d11
CAS 1189985-02-1
Renhedsgrad noter HPLC
Molekylær formel C23 D11 H17 Cl N3 O5 S
Mængde 10 mg
Synonym Benzamide, 5-chloro-N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxy- (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxybenzamide (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexyl-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxybenzamide, Glibenclamide-d11, Glibenclamide-D11 (cyclohexyl-D11), Glyburide-d11 (cyclohexyl-d11)
InChI formel InChI=1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)/i2D2,3D2,4D2,5D2,6D2,18D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(NC(=O)NS(=O)(=O)c2ccc(CCNC(=O)c3cc(Cl)ccc3OC)cc2)C([2H])([2H])C1([2H])[2H]
IUPAC navn 5-chloro-2-methoxy-N-[2-[4-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]benzamide
Molekylvægt (g/mol) 505.071
Formel vægt 504.213
Procent renhed >95