Få mere at vide
Glyburide-d11 (cyclohexyl-d11), CDN
Artikelnummer. 30320521
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320521 0.005 g
30309238 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320521 Leverandør CDN Leverandørnr. CDND78880.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glyburide-d11
CAS 1189985-02-1
Molekylær formel C23 D11 H17 Cl N3 O5 S
Mængde 0.005 g
Synonym Benzamide, 5-chloro-N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxy- (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxybenzamide (ACI), 5-Chloro-N-[2-[4-[[[(cyclohexyl-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxybenzamide, Glibenclamide-d11, Glibenclamide-D11 (cyclohexyl-D11), Glyburide-d11 (cyclohexyl-d11)
InChI formel InChI=1 S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18 H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)/i2D2,3D2,4D2,5D2,6D2,18 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(NC(=O)NS(=O)(=O)c2ccc(CCNC(=O)c3cc(Cl)ccc3OC)cc2)C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 5-chloro-2-methoxy-N-[2-[4-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]benzamide
Molekylvægt (g/mol) 505.071
Formel vægt 504.213 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity