missing translation for 'onlineSavingsMsg'
Få mere at vide

MedChemExpress Gly-Arg-Gly-Asp-Ser

Artikelnummer. 16490396
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 mg
Pakningsstørrelse:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16490396 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16490396 Leverandør MedChemExpress Leverandørnr. HYP0295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gly-Arg-Gly-Asp-Ser is a pentapeptide that forms the cell-binding domain of a glycoprotein, osteopontin. Gly-Arg-Gly-Asp-Ser binds to integrin receptors αvβ3 and αvβ5 with estimated IC50 of ∼5 and ∼6.5 μM

Specifications

Renhed 95.05%
CAS 96426-21-0
Indhold og opbevaring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molekylvægt (g/mol) 490.47 g/mol
Produkttype Peptides
Mængde 2 mg
Forsendelsestilstand Room temperature
Opløselighed DMSO : 50 mg/mL (101.94 mM∣Need ultrasonic) ∣H2O : 33.33 mg/mL (67.96 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.