Få mere at vide
Glutaronitrile-d6, TRC
Artikelnummer. 30403960
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30403960 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403960 Leverandør Toronto Research Chemicals Leverandørnr. TRCG598103100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glutaronitrile-d6
CAS 1346598-53-5
Molekylær formel C5D6N2
Mængde 100 mg
Synonym 1,3-Dicyanopropane-d6, 1,5-Pentanedinitrile-d6, Glutaric Acid Dinitrile-d6, Glutaric Dinitrile-d6, Glutarodinitrile-d6, NSC 3807-d6
InChI formel InChI=1S/C5H6N2/c6-4-2-1-3-5-7/h1-3H2/i1D2,2D2,3D2
SMIL [2H]C([2H])(C#N)C([2H])([2H])C([2H])([2H])C#N
IUPAC navn 2,2,3,3,4,4-hexadeuteriopentanedinitrile
Molekylvægt (g/mol) 100.15
Formel vægt 100.0908