Få mere at vide
Glutaric-d4 Acid, TRC
Artikelnummer. 30408552
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30408552 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408552 Leverandør Toronto Research Chemicals Leverandørnr. TRCG598085500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glutaric-d4 Acid
CAS 19136-99-3
Molekylær formel C5 D4 H4 O4
Mængde 500 mg
Synonym Pentanedioic-2,2,4,4-d4 Acid, Pentanedioic-2,2,4,4-d4 acid (9CI), Glutaric-2,2,4,4-d4 acid (8CI), Pentanedioic acid-2,2,4,4-d4, Glutaric acid-2,2,4,4-d4, Glutaric acid-d4, Pentanedioic-d4 acid
InChI formel InChI=1S/C5H8O4/c6-4(7)2-1-3-5(8)9/h1-3H2,(H,6,7)(H,8,9)/i2D2,3D2
SMIL [2H]C([2H])(CC([2H])([2H])C(=O)O)C(=O)O
IUPAC navn 2,2,4,4-tetradeuteriopentanedioic acid
Molekylvægt (g/mol) 136.14
Formel vægt 136.0674