Learn More
Glutaric anhydride, 95%, Thermo Scientific Chemicals
Catalog Number p-138410
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 46.0°C to 57.0°C
Color White
Boiling Point 290.0°C
Flash Point 173°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, V,11, 9
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in benzene, toluene, acetone and, chloroform
Formula Weight 114.1
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Glutaric anhydride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glutaric anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.