missing translation for 'onlineSavingsMsg'
Learn More

Glutaric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-138399
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
110-94-1
C5H8O4
132.12
MFCD00004410
JFCQEDHGNNZCLN-UHFFFAOYSA-N
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
743
CHEBI:17859
pentanedioic acid
OC(=O)CCCC(O)=O
This item is not returnable. View return policy
110-94-1
C5H8O4
132.12
MFCD00004410
JFCQEDHGNNZCLN-UHFFFAOYSA-N
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
743
CHEBI:17859
pentanedioic acid
OC(=O)CCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 92.0°C to 99.0°C
Color Orange
Boiling Point 302.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HO2C(CH2)3CO2H
Quantity 25 g
Beilstein 02, 631
Fieser 01,412
Merck Index 15, 4509
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: freely soluble in abs. alcohol and ether, soluble in benzene and chloroform
Formula Weight 132.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glutaric acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'