missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Glutamine Synthetase Antibody (GLUL/8996R), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24031AF532
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| Glutamine Synthetase | |
| Monoclonal | |
| Alexa Fluor 532 | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| Recombinant fragment (around aa 200-373) of human Glutamine Synthetase protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 2752 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| GLUL/8996R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion