Få mere at vide
Novus Biologicals™ Glut1 Recombinant Protein Antigen
Artikelnummer. 18296495 Shop alle Bio Techne produkter
Skift visning
Protein længde:
GRTFDEIASGFRQGGASQSDKTPEELFHP
RNEENRAKSVLKKLRGTADVTHDLQE
Pakningsstørrelse:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde unitSize
18296495 RNEENRAKSVLKKLRGTADVTHDLQE -
18136537 GRTFDEIASGFRQGGASQSDKTPEELFHP 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18296495 Leverandør Novus Biologicals™ Leverandørnr. NBP254745PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Glut1 The Glut1 Recombinant Protein Antigen is derived from E. coli. The Glut1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Protein længde RNEENRAKSVLKKLRGTADVTHDLQE
Oprensningsmetode >80% by SDS-PAGE and Coomassie blue staining
Fælles navn Glut1 Recombinant Protein Antigen
Indhold og opbevaring Store at −20°C. Avoid freeze-thaw cycles.
Formulering PBS (pH 7.4), 1 M Urea
Til brug med (applikation) Blocking/Neutralizing, Control
Gene Alias DYT17, DYT18, DYT18DYT17, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, Glut1
Gen symbol SLC2A1
Etikettype Unlabeled
Produkttype Recombinant Protein Antigen
Mængde 0.1mL
Regulatorisk status RUO
Forskningskategori Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.