missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1 Monoclonal Antibody (RH95), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518117
Additional Details : Weight : 0.01000kg
Description
This antibody contains enough material to conduct 10 mini-Western blots.
GluR1 (glutamate receptor 1) is an ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter, L-glutamate, induces a conformation change that leads to the opening of the cation channel and converts the chemical signal to an electrical impulse. Then, the receptor desensitizes rapidly and enters a transiet inactive state characterized by the presence of a bound agonist. In the presence of CACNG4, CACNG7, or CACNG8 GluR1 shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.Specifications
GluR1 | |
Monoclonal | |
∽1.0 mg/mL | |
0.01M HEPES with 0.15M NaCl, 0.1mg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P19490, P23818 | |
GRIA1 | |
Synthetic peptide corresponding to the N-terminal region of rat GRIA1 conjugated to KLH | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG2a |
Western Blot | |
RH95 | |
Unconjugated | |
GRIA1 | |
2900051M01Rik; AI853806; AMPA 1; AMPA glutamate receptor; AMPA receptor GluR1/A; AMPA-selective glutamate receptor 1; Glr1; Glr-1; GluA1; GLUH 1; GLUH1; GluR K1; GLUR1; Glur-1; GluR1 flip; GluR1 flop; GLURA; GluR-A; GluRK1; gluR-K1; glutamate ionotropic receptor AMPA type subunit 1; glutamate receptor; glutamate receptor 1; glutamate receptor A; glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; Gria1; GRM1A; HBGR1; HGBR1; HIPA; HIPA1; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1 | |
Mouse | |
Protein G | |
RUO | |
14799, 50592 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |