Få mere at vide
MedChemExpress γ-Glu-Phe
Artikelnummer. 16494586
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16494586 100 mg
1 options
Artikelnummer. 16494586 Leverandør MedChemExpress Leverandørnr. HY101399
MedChemExpress γ-Glu-Phe (γ-Glutamylphenylalanine) is synthesized by Bacillus amyloliquefaciens (GBA) and Aspergillus oryzae (GAO). γ-Glu-Phe or the post-enzymatic reaction mixture enhances the umami intensity of commercial soy sauce and model chicken broth.
Tekniske data
Renhed 99.85%
CAS 7432-24-8
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molekylvægt (g/mol) 294.3 g/mol
Produkttype Natural Products
Mængde 100 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : ≥ 50 mg/mL (169.89 mM)
Synonym γ-Glutamylphenylalanine