missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
GLS2 Antibody (CL7716) [DyLight 488], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44209G
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GLS2 Monoclonal antibody specifically detects GLS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| GLS2 | |
| Monoclonal | |
| DyLight 488 | |
| EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2 | |
| This antibody was generated using a synthetic peptide of Q9UI32, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 27165 | |
| Store at 4°C in the dark. | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CL7716 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion