Få mere at vide
GLPG0492 (R enantiomer), MedChemExpress
Artikelnummer. 16469947
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16469947 5 mg
16457468 10 mg
16427436 100 mg
16403497 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16469947 Leverandør Medchem Express Leverandørnr. HY18102A
MedChemExpress GLPG0492 R enantiomer is the R enantiomer of GLPG-0492, which is a novel selective androgen receptor modulator.
GLPG0492 R enantiomer is the R enantiomer of GLPG-0492, which is a novel selective androgen receptor modulator.
Tekniske data
Kemisk navn eller materiale GLPG0492 (R enantiomer)
Fysisk form Solid
CAS 1215085-93-0
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C19H14F3N3O3
Til brug med (applikation) Neuroscience-Neuromodulation
Opløselighedsinformation DMSO : 100 mg/mL (256.85 mM; Need ultrasonic)
SMIL N#CC1=CC=C(N(C(N(C)[C@]2(C3=CC=CC=C3)CO)=O)C2=O)C=C1C(F)(F)F
Molekylvægt (g/mol) 389.33
Formel vægt 389.33
Procent renhed 97.35%