missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress GLP-2(1-33)(human)

Product Code. 16412008
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16412008 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412008 Supplier MedChemExpress Supplier No. HYP1024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLP-2(1-33) (human) is an enteroendocrine hormone which can bind to the GLP-2 receptor and stimulate the growth of intestinal epithelium.

Specifications

Renhed 99.18%
CAS 223460-79-5
Indhold og opbevaring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Molekylvægt (g/mol) 3766.19 g/mol
Produkttype Peptides
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 25 mg/mL (6.64 mM∣ultrasonic and adjust pH to 9 with NH4OH)
Synonym GLP-2 (human)∣ Glucagon-like peptide 2 (human)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.