Få mere at vide
Invitrogen™ GLP-1 Monoclonal Antibody (03)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ ABS0460302
Produktkode 15366291
  • 6170.00 DKK / 400µL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Specificity: ABS 046-03-02 binds GLP-1 (1-37), GLP-1 (7-37) and GLP-1 (9-37). This antibody shows 0-16% cross-Specificity with C-terminally amidated GLP-1 in various circumstances. Reactivity: ABS 046-03-02 binds free GLP-1 (1-37), GLP-1 (7-37) and GLP-1 (9-37) in solution. Two different ELISA sandwich setups can be used for measuring non-amidated GLP-1 forms: The first uses ABS 046-03 as a capture antibody with biotinylated detection antibody ABS 033-10-02. This shows ~5% cross-reactivity with C-terminally amidated GLP-1. The second combination uses HYB 147-12 as a capture antibody with biotinylated detection antibody ABS 046-03-02, resulting in ~16% cross-reactivity with C-terminally amidated GLP-1. Results show detection limits of 44 pmol/L which is 10-20 times higher than the basal concentration of GLP-1, so the assays have to be optimized. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
GLP-1
Monoclonal
1.03 mg/mL
PBS with 15mM sodium azide; pH 7.4
P01275, P06883, P55095
GCG
Synthetic GLP-1(9-37) adsorbed onto aluminum hydroxide gel.
400 μL
Primary
Human, Mouse, Rat, Mammalia
Antibody
IgG1 κ
ELISA, Western Blot
03
Unconjugated
GCG
GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon
Mouse
Protein A
RUO
14526, 24952, 2641
4°C, store in dark
Liquid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner