Få mere at vide
SICCO Glove Box Antistatic with Transfer Chamber
Artikelnummer. 15835931
Dimensioner (L x B x H):
1210 x 780 x 600 mm
Artikelnummer. 15835931
Brand: SICCO V199208
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Glove Box Antistatic with Transfer Chamber POLYCARBONATE/ALUMINIUM base made of PE. Usable interior space: W 850 x H 650 x D 540 mm
  • Aluminium frame with static dissipative panels made of polycarbonate, base panel made of static dissipative polyethylene.
  • Front panel with two glove ports and one pair of static dissipative EPDM gloves size 9,75, two lead-ins for cable on the back side (upper left and upper right).
  • Transfer chamber with two doors, external door with latch to prevent accidentally opening, the tray extends into the workspace with opening of the internal door for easy removal of inserted products.
  • Work space and transfer chamber each including two hose connectors for gas-filling and aeration and appropriate tubing (2x 5 m tubing for gas-filling made of PVC, 2x 5 m aeration tubing made of PE).
  • All materials used can discharge electrostatic charging by means of a grounding cable which can be connected on the back side (connecting thread M6).
  • Robust and light-weight aluminium construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
  • Transfer chamber for inserting further products
Tekniske data
Produkttype Glove Box
Dimensioner (L x B x H) 1210 x 780 x 600 mm
Volumen (metrisk) 290 L
Vægt (metrisk) 20 kg