Få mere at vide
STEMCELL Technologies GloCell™ Fixable Viability Dye Violet 540
Artikelnummer. 17160362
Encompass_Preferred
Skift visning
Mængde:
100 Tests
5 x 100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17160362 100 Tests
17119291 5 x 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17160362 Leverandør STEMCELL Technologies Leverandørnr. 75011.1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Amine-labeling fluorescent dye for live/dead staining of mammalian cells
  • GloCell™ Fixable Viability Dye Violet 540 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
  • The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
  • Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
  • GloCell™ Fixable Viability Dye Violet 540 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 540 nm that can be detected using the 525/50 band pass filter used to detect AmCyan
Tekniske data
Til brug med (applikation) Flow Cytometry; Immunofluorescence
Mængde 100 Tests