Få mere at vide
STEMCELL Technologies GloCell™ Fixable Viability Dye Violet 510
Beskrivelse
- GloCell™ Fixable Viability Dye Violet 510 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
- The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
- Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
- GloCell™ Fixable Viability Dye Violet 510 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 510 nm that can be detected using the 525/50 band pass filter used to detect AmCyan
Tekniske data
Tekniske data
| Til brug med (applikation) | Flow Cytometry; Immunofluorescence |
| Mængde | 5 x 100 Tests |