Få mere at vide
STEMCELL Technologies GloCell™ Fixable Viability Dye Red 710
Artikelnummer. 17120362
Encompass_Preferred
Skift visning
Mængde:
100 Tests
5 x 100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17120362 100 Tests
17169281 5 x 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17120362 Leverandør STEMCELL Technologies Leverandørnr. 75006.1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Amine-labeling fluorescent dye for live/dead staining of mammalian cells
  • GloCell™ Fixable Viability Dye Red 710 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
  • The dye irreversibly binds intracellular and cell surface amine groups and can be used prior to fixation, permeabilization, or cryopreservation
  • Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in greater fluorescence compared to live cells in a cell viability assay
  • GloCell™ Fixable Viability Dye Red 710 is excited by the red laser (633 nm) with an excitation maximum of 647 nm, and it has a fluorescence emission maximum of 710 nm that can be detected using the 710/50 band pass filter used to detect Alexa Fluor™ 700
Tekniske data
Til brug med (applikation) Flow Cytometry; Immunofluorescence
Mængde 100 Tests