Beskrivelse
- Non-sterile
- Molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents
- Polystyrene tubes can withstand 3000rcf
- Polypropylene tubes can withstand 5000rcf and are autoclavable and freezable
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 3 mL |
| Længde (metrisk) | 55 mm |
| Materiale | Polystyrene |
| Emballage | 1000/Cs. |
| Kantet | No |
| Sterilitet | Non-sterile |
| Autoklaverbar | Not Autoclavable |
| Uddannet | No |
| Diameter (metrisk) | 12 mm |
| Produkttype | Test Tube |