Learn More
Brady™ GlobalMark™ Four Color Labeling Ribbon With Panel
This super-tough smear-proof GlobalMark™ Four Color Labeling Ribbon With Panel assures your printed text lasts for years outdoors and in an industrial environment.
3205.00 DKK
Specifications
Product Type | Printer Ribbon |
---|---|
Color | Black/Blue/Red/Yellow |
For Use With (Equipment) | GlobalMark Printer |
Length (English) | 65.6 yd. |
Width (English) | 4.13 in. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11539313
|
Brady™
076791 |
105 mm Black/Red/Blue/Yellow Ribbon, 200 mm Panel Length |
3205.00 DKK
1 box |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11529313
|
Brady™
076790 |
105 mm Black/Red/Blue/Yellow Ribbon, 400 mm Panel Length |
3205.00 DKK
1 box |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- Ribbons ship self-contained in cartridges that drop straight in to the GlobalMark™ printer with a ''click''.
- Patent D481,7555.
Specifications
Printer Ribbon | |
GlobalMark Printer | |
4.13 in. | |
105 mm | |
Printed text last for years outdoors and in industrial areas |
Black/Blue/Red/Yellow | |
65.6 yd. | |
60 m | |
60 m x 105 mm | |
1 Ea. |