Få mere at vide
Glipizide EP Impurity F, Mikromol™
Artikelnummer. 30051488
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051488 100 mg
30049694 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051488 Leverandør Mikromol Leverandørnr. MM0942.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Glipizide EP Impurity F
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl [2-(4-Sulphamoylphenyl)ethyl]carbamate
CAS 192118-08-4
Analyt- eller komponentnavne Ethyl [2-(4-Sulphamoylphenyl)ethyl]carbamate
Forsendelsestilstand Room Temperature
Molekylær formel C11 H16 N2 O4 S
Certificeringer/compliance ISO 9001
Synonym Ethyl [2-(4-Sulphamoylphenyl)ethyl]carbamate, Glipizide Imp. F (EP), Glipizide Impurity F
InChI formel InChI=1S/C11H16N2O4S/c1-2-17-11(14)13-8-7-9-3-5-10(6-4-9)18(12,15)16/h3-6H,2,7-8H2,1H3,(H,13,14)(H2,12,15,16)
SMIL CCOC(=O)NCCc1ccc(cc1)S(=O)(=O)N
IUPAC navn ethyl N-[2-(4-sulfamoylphenyl)ethyl]carbamate
Molekylvægt (g/mol) 272.32
Mængde 100 mg
Formel vægt 272.0831 g/mol