Få mere at vide
Glipizide-d11, TRC
Artikelnummer. 30403312
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30403312 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403312 Leverandør Toronto Research Chemicals Leverandørnr. TRCG41022710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glipizide-d11
CAS 1189426-07-0
Renhedsgrad noter HPLC
Molekylær formel C21 D11 H16 N5 O4 S
Mængde 10 mg
Synonym 2-Pyrazinecarboxamide, N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl- (ACI), N-[2-[4-[[[(Cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide (ACI), Glipizide D11 (cyclohexyl D11)
InChI formel InChI=1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)/i2D2,3D2,4D2,5D2,6D2,17D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(NC(=O)NS(=O)(=O)c2ccc(CCNC(=O)c3cnc(C)cn3)cc2)C([2H])([2H])C1([2H])[2H]
IUPAC navn 5-methyl-N-[2-[4-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]pyrazine-2-carboxamide
Molekylvægt (g/mol) 456.603
Formel vægt 456.247
Procent renhed >95