Få mere at vide
Gliclazide Impurity D, TRC
Artikelnummer. 17934115
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17934115 50 mg
17920555 10 mg
17960875 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17934115 Leverandør Toronto Research Chemicals Leverandørnr. TRCG40988550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1136426-19-1
Molekylær formel C15 H20 N2 O3 S
Molekylvægt (g/mol) 308.3959
Synonym Cyclopenta[c]pyrrole-2(1H)-carboxamide, hexahydro-N-[(4-methylphenyl)sulfonyl]-, N-[(4-Methylphenyl)sulfonyl]hexahydrocyclopenta[c]pyrrol-2(1H)-carboxamide, Gliclazide Imp. D (EP), Hexahydro-N-[(4-methylphenyl)sulfonyl]-cyclopenta[c]pyrrole-2(1H)-carboxamide
IUPAC navn N-(4-methylphenyl)sulfonyl-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxamide
SMIL Cc1ccc(cc1)S(=O)(=O)NC(=O)N2CC3CCCC3C2
Tekniske data
Kemisk navn eller materiale Hexahydro-N-[(4-methylphenyl)sulfonyl]-cyclopenta[c]pyrrole-2(1H)-carboxamide
Mængde 50 mg
InChI formel InChI=1S/C15H20N2O3S/c1-11-5-7-14(8-6-11)21(19,20)16-15(18)17-9-12-3-2-4-13(12)10-17/h5-8,12-13H,2-4,9-10H2,1H3,(H,16,18)
Formel vægt 308.1195