Learn More
Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals
Widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.
1736.33 DKK - 5186.64 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334307
|
Thermo Scientific Alfa Aesar
037997.DJ |
50 mm |
1736.33 DKK
50mm |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11324307
|
Thermo Scientific Alfa Aesar
037997.DM |
100 mm |
3077.70 DKK
100mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15491467
|
Thermo Scientific Alfa Aesar
037997.DR |
200 mm |
5186.64 DKK
200mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Glassy carbon rod, type 2 is widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.