Learn More
Glassy carbon plate, 3mm (0.1in) thick, type 1, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
1095.45 DKK - 10388.18 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390008
|
Thermo Scientific Alfa Aesar
042821.FF |
25 x 25 mm |
1095.45 DKK
25mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11356159
|
Thermo Scientific Alfa Aesar
042821.FI |
50 x 50 mm |
3912.33 DKK
50mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11380008
|
Thermo Scientific Alfa Aesar
042821.GH |
100 x 100 mm |
10388.18 DKK
100mm |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.