Learn More
Glassy carbon plate, 3mm (0.1in) thick, type 1, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
1095.45 DKK - 10388.18 DKK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11390008
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042821.FF
25 x 25 mm
1095.45 DKK
25mm
View alternative products
Save Up to 
»
In Stock
to see stock.
11356159
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042821.FI
50 x 50 mm
3912.33 DKK
50mm
View alternative products
Save Up to 
»
In Stock
to see stock.
11380008
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042821.GH
100 x 100 mm
10388.18 DKK
100mm
View alternative products
Save Up to 
»
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Videos
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only