Learn More
Glassy carbon plate, 3mm (0.1 in.) thick, type 1, Alfa Aesar™
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
Brand: Alfa Aesar 042821.FI
Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg
Specifications
Glassy carbon plate | |
50 x 50mm | |
MFCD00133992 | |
≈2.9g/25x25mm |
Tube | |
7440-44-0 | |
Insoluble. |